missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC12A3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA555969

Catalog No. PIPA555969


Only null left
Explore available promotions

Description

Description

Immunogen sequence: LLIPYLLGRK RRWSKCKIRV FVGGQINRMD QERKAIISLL SKFRLGFHEV HILPDINQNP RAEHTKRFED MIAPFRLNDG FKDEATVNEM RRDCPWKISD EEITKNRVKS LRQVRLNEIV LDYSRDAALI VITLPIGRKG KCPSS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

The SLC12A3 gene encodes the thiazide-sensitive sodium-chloride symporter (NCC), which is critical for salt balance and blood pressure regulation in humans. This symporter is predominantly expressed in the distal convoluted tubule of the kidney, where it facilitates the reabsorption of sodium and chloride ions from the renal filtrate back into the bloodstream. NCC activity plays a vital role in maintaining extracellular fluid volume and potassium homeostasis, impacting systemic blood pressure. Mutations in SLC12A3 can lead to Gitelman syndrome, a hereditary salt-wasting disorder characterized by hypokalemic metabolic alkalosis, low blood pressure, and hypomagnesemia. This gene is regulated by the renin-angiotensin system, which modulates NCC activity through phosphorylation mediated by WNK kinases, thereby affecting renal sodium handling and blood pressure modulation. Research highlights the therapeutic potential of targeting NCC with thiazide diuretics to treat hypertension, emphasizing its importance in both physiological and pathological contexts. The dynamic regulation of NCC by hormonal pathways underscores its significance in fluid and electrolyte balance, contributing to cardiovascular health.
Specifications

Specifications

SLC12A3
Polyclonal
Unconjugated
Slc12a3
AI035291; chemokine (C-C); CKb10; CK-beta-10; MCP-4; MGC17134; Na-Cl cotransporter; NaCl electroneutral thiazide-sensitive cotransporter; Na-Cl symporter; Ncc; NCC1; NCC-1; NCCT; SCYA13; SCYL1; si:ch211-122c9.4; SLC12A3; sodium chloride cotransporter; solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; thiazide-sensitive sodium chloride cotransporter; thiazide-sensitive sodium-chloride cotransporter; TSC; wu:fj83c03
Rabbit
Antigen affinity chromatography
RUO
6559
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P55017
Slc12a3
Recombinant protein corresponding to Human SLC12A3. Recombinant protein control fragment (Product #RP-102376).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions