missing translation for 'onlineSavingsMsg'
Learn More

Phytosterols, Practical Grade, MP Biomedicals™

$82.17 - $82.17

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
Molecular Weight (g/mol) 414.718
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym 22,23-dihydrostigmasterol, α-Dihydrofucosterol, 24β-Ethylcholesterol, 5-Stigmasten-3β-ol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10288680
View Documents
MP Biomedicals Inc
0210288680
100 g
Each for $82.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

83-46-5
414.718
22,23-dihydrostigmasterol, α-Dihydrofucosterol, 24β-Ethylcholesterol, 5-Stigmasten-3β-ol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
CHEBI:27693
CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
C29H50O
KZJWDPNRJALLNS-VJSFXXLFSA-N
222284
(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol

Specifications

83-46-5
C29H50O
22,23-dihydrostigmasterol, α-Dihydrofucosterol, 24β-Ethylcholesterol, 5-Stigmasten-3β-ol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
414.718
CHEBI:27693
Practical
Phytosterols
White
100 g
KZJWDPNRJALLNS-VJSFXXLFSA-N
(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
222284
414.7
Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'