missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ sDIBO Alkyne Kits for Antibody Labeling

Catalog No. S10911
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Biotin
pHrodo Red
Primary Amine
Labeling Scale:
100-250 μg
1 mg
5 mg
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Labeling Scale
S10911 Alexa Fluor 647 5 mg
C20027 Alexa Fluor 488 100-250 μg
C20029 Alexa Fluor 647 100-250 μg
C20031 Primary Amine 100-250 μg
C20034 pHrodo Red 100-250 μg
S10902 Biotin 1 mg
S10904 Alexa Fluor 488 1 mg
S10907 Biotin 5 mg
C20028 Alexa Fluor 555 100-250 μg
C20030 Biotin 100-250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. S10911 Supplier Invitrogen™ Supplier No. S10911

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.

Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 μg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 μg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

Specifications

Quantity for 5 mg of Ab
Kit Contents Contains 1 vial of SiteClick Alexa Fluor 647 sDIBO Alkyne for Antibody Labeling in 250 μL DMSO and 1 antibody concentrator
Filter Type Cy5
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Chemical Reactivity Azide
Flow Cytometer Laser Lines Red
Labeling Method Click Chemistry
Labeling Scale 5 mg
Shipping Condition Wet Ice
Reactive Moiety sDIBO Alkyne
Label or Dye Alexa Fluor 647
Color Far-red
Product Line Alexa Fluor, SiteClick
Labeling Target Azido modified antibodies
Excitation/Emission 650/665 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.