missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Duet and Quartet Antibody Labeling Kits

Catalog No. S11003
Change view
Click to view available options
Product Type:
Duet Antibody Modification Kit
Quartet Antibody Modification Kit
Quantity:
100 μg
3 x 100 μg
1 mg
5 mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
S11003 Quartet Antibody Modification Kit 100 μg
S11001 Quartet Antibody Modification Kit 5 mg
S11002 Quartet Antibody Modification Kit 1 mg
S11004 Duet Antibody Modification Kit 5 mg
S11005 Duet Antibody Modification Kit 1 mg
S11006 Duet Antibody Modification Kit 100 μg
S11007 Quartet Antibody Modification Kit 3 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S11003 Supplier Invitrogen™ Supplier No. S11003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Introducing SiteClick Duet and Quartet kits. Experience the same robust, covalent, site-specific antibody conjugation you trust, now with a faster and easier workflow. Complete in less than 24 hours with fewer workflow steps.

SiteClick Duet and Quartet antibody labeling kits are used to create label-ready, site-specific, azido-modified antibodies without lengthy genetic modifications. Our enzyme-driven SiteClick labeling precisely attaches an azido moiety to the heavy chains of IgG antibodies, preserving antigen-binding domains for optimal target binding. This site selectivity targets carbohydrate domains on the Fc region, applicable to all IgG antibodies regardless of isotype or host species.

The SiteClick Quartet Antibody Labeling Kit offers up to four payloads per antibody (DOS 4) in less than 24 hours, while the SiteClick Duet Antibody Labeling Kit enables the attachment of two payloads (DOS 2) in only 4 hours.

  • Features of the SiteClick Duet and Quartet antibody labeling kits include:
    • Modify any IgG antibody species or isotype with a complete kit
    • Follow a brief, straightforward protocol
    • Preserve antibody function with efficient, site-specific labeling
    • Achieve consistent results with reproducible labeling chemistry
    • Develop therapeutic antibodies, antibody-drug conjugates, antibody-oligo conjugates, bispecific antibodies, and immunodiagnostics
  • Once azido-modified, choose from a variety of SiteClick sDIBO or DBCO alkyne labels for covalent attachment via copper-free click chemistry under mild conditions.
  • Enjoy the flexibility to select different fluorescent labels, including Alexa Fluor, pHrodo, and LysoLight dyes, or to conjugate other molecules such as biotin, oligos, toxins, or degraders.
  • SiteClick labeling is useful for developing therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostic tests, helping ensure high specificity, reproducible labeling, and preserved antibody performance.
  • Unlock the full potential of your antibodies with our Custom SiteClick Antibody Labeling Service and customized labels.
  • If conventional methods have failed or reduced your antibody's activity, our expert team can help.
  • We offer SiteClick-compatible labels in bulk format, and custom labeling with a variety of detection molecules, including Alexa Fluor, LysoLight, and pHrodo dyes, as well as biotin and R-PE.
  • Interested in using SiteClick technology beyond your internal research? Let us help you use this in your product manufacturing and services.
  • Please inquire about licensing options for your commercial applications.

Specifications

For Use With (Application) Antibody Production and Labeling
Product Type Quartet Antibody Modification Kit
Quantity 100 μg
Sufficient For 1 Labeling
Content And Storage 2°C to 8°C
For Use With (Equipment) Benchtop Centrifuge
Labeling Method Click Chemistry
Shipping Condition Wet Ice
Product Line SiteClick
Reactive Moiety Azide
Format Labeling Kit
Labeling Target IgG, Antibodies
Species Any
Label Type Degree of Labeling: up to 4
Incubation Time 16 to 20 hr.
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.