missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Qdot™ Antibody Labeling Kits

Catalog No. p-7222417
Encompass
Change view
Click to view available options
Label or Dye:
Qdot 525
Qdot 585
Qdot 605
Qdot 625
Qdot 655
Qdot 705
Qdot 800
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
S10453 Qdot 655
S10449 Qdot 525
S10454 Qdot 705
S10451 Qdot 585
S10452 Qdot 625
S10455 Qdot 800
S10469A Qdot 605
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S10453 Supplier Invitrogen™ Supplier No. S10453
Only null left

Achieve precise and reproducible antibody labeling with SiteClick R-PE Antibody Labeling Kit.

Achieve precise and reproducible antibody labeling with SiteClick R-PE Antibody Labeling Kit. This kit targets carbohydrate domains present on all IgG antibodies, regardless of isotype or host species, ensuring site-selective labeling. The labeled antibodies are suitable for flow cytometry, fluorescence imaging, and western blot detection.

Features of the SiteClick R-PE Antibody Labeling Kit include:

  • Complete kit—includes everything needed to label 100 μg of IgG antibody using a straightforward, step-by-step protocol
  • Site-specific labeling—SiteClick technology attaches labels to the heavy chains of IgG antibodies under mild conditions, preserving antigen-binding domains
  • Consistent results—the gentle, site-selective attachment to heavy-chain N-linked glycans ensures reproducibility across sequential labelings and different antibodies, without harsh reduction steps
  • Efficient protocol—easy-to-follow steps requiring minimal hands-on time
  • High-quality conjugates—produces highly efficient, site-specific, and reproducible antibody conjugates

Unlike traditional amine or thiol labeling, which can affect antigen binding domains or antibody structure, SiteClick labeling technology maintains the integrity of the antibody. This ensures that antigen binding sites remain available for target binding, providing consistent and reproducible labeling. The workflow utilizes copper-free click chemistry to covalently link the DIBO moiety with the azide-modified antibody, ensuring uniform labeling without the need for reaction optimization.

With the SiteClick R-PE Antibody Labeling Kit, you can confidently achieve consistent, high-quality labeling every time.

Specifications

Kit Contents Contains everything needed to azido modify and label 100 μg of antibody.
Product Type Labeling Kit
Quantity 1 kit
Content And Storage Store the entire kit at 2 to 6°C upon receipt
Excitation/Emission 405/655
Labeling Method SiteClick Labeling System
Labeling Scale 100 μg
Shipping Condition Wet Ice
Product Line Qdot, SiteClick
Label or Dye Qdot 655
Color Far-Red
Labeling Target IgG
Filter Type Qdot 525-800
Flow Cytometer Laser Lines UV, Violet, Blue, Green, Yellow
Show More Show Less