missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Antibody Azido Modification Kit

Catalog No. S20026
Encompass
Change view
Click to view available options
Labeling Scale:
100 to 250 μg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Labeling Scale
S20026 100 to 250 μg
S10901 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S20026 Supplier Invitrogen™ Supplier No. S20026

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Create a label-ready site-specific azido-modified antibody without lengthy genetic modification using the SiteClick Antibody Azido Modification Kit.

Create a label-ready, site-specific, azido-modified antibody without lengthy genetic modification using the SiteClick Antibody Azido Modification Kit. Use this kit to carry out a two-step conjugation process over three days. SiteClick labeling uses enzymes to attach an azido moiety to the heavy chains of an IgG antibody, ensuring the antigen-binding domains remain unaltered.

Create a label-ready, site-specific, azido-modified antibody without lengthy and inefficient genetic modification using the SiteClick Antibody Azido Modification Kit. SiteClick labeling uses enzymes to specifically attach an azido moiety to the heavy chains of an IgG antibody, ensuring that the antigen-binding domains remain unaltered for binding to the antigen target. This site selectivity is achieved by targeting the carbohydrate domains present on the Fc region of essentially all IgG antibodies regardless of isotype and host species.

Once azido–modified, a variety of SiteClick sDIBO or DBCO alkyne labels are available for covalent attachment to the antibody via copper-free click chemistry under mild reaction conditions at your choice of scale. This provides the flexibility to choose different fluorescent labels for your antibody, attach another molecule via streptavidin, or attach your own molecule, such as an oligo, drug, or toxin via amine-reactive or amine-containing moieties depending on your assay. This site-specific modification using SiteClick labeling is a critical tool in the development of therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostic tests that require high specificity with reproducible labeling reactions and helps ensure that antibody performance and function are preserved after conjugation.

Features of the SiteClick Antibody Azido Modification Kit include:
• Contains everything required to azido-modify any species or isotype of IgG antibody
• Easy-to-follow step-by-step protocol
• Highly efficient, site-specific labeling preserves antibody function and results in high quality antibody conjugate
• Reproducible labeling chemistry with lot-to-lot consistency in degree of labeling
• Optimal method for site-specific labeling of therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostics

Custom SiteClick Antibody Labeling Service and sDIBO labels
If you have an antibody that is considered 'difficult to label' or has lost activity after labeling using a conventional method, please contact our custom service representatives to determine whether the SiteClick Antibody Labeling Service would be right for your antibody. We offer complete custom SiteClick antibody labeling services with the option of multiple detection molecules including biotin, Alexa Fluor dyes, Qdot fluorophores, R-PE, chelates for PET imaging, and many others.

Order Info

Store in refrigerator (2–8°C), desiccate, protect from light, and do not freeze.

Specifications

Product Type Antibody Azido Modification Kit
Quantity for 100-250 μg of Ab
Content And Storage Store in refrigerator (2–8°C), desiccate, protect from light, and do not freeze.

Labeling Scale 100 to 250 μg
Shipping Condition Room Temperature
Product Line SiteClick
Chemical Reactivity Azide
Label or Dye Azide, Alexa Fluor 555, Alexa Fluor 488
Color Clear
Labeling Target Antibodies

For Research Use Only. Not for use in diagnostic procedures.