missing translation for 'onlineSavingsMsg'
Learn More

Sitafloxacin sesquihydrate, Thermo Scientific Chemicals

Catalog No. AC464372500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464372500 250 mg
AC464370010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464372500 Supplier Thermo Scientific Chemicals Supplier No. 464372500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163253-35-8
Molecular Formula C38H42Cl2F4N6O9
Molecular Weight (g/mol) 873.68
InChI Key MPORYQCGWFQFLA-UHFFFAOYNA-N
IUPAC Name bis(7-{7-amino-5-azaspiro[2.4]heptan-5-yl}-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid) trihydrate
SMILES O.O.O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 436.84
Chemical Name or Material Sitafloxacin sesquihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only