accessibility menu, dialog, popup

UserName

NH2NClNOOOHFFNH2NClNOOOHFFH2OH2OH2O

CAS RN 163253-35-8

IUPAC Name: bis(7-{7-amino-5-azaspiro[2.4]heptan-5-yl}-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid) trihydrate
Synonyms: bis(7-{7-amino-5-azaspiro[2.4]heptan-5-yl}-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-4-oxoquinoline-3-carboxylic acid) trihydrate
Molecular Weight (g/mol): 873.68
Molecular Formula: C38H42Cl2F4N6O9
InChi Key: MPORYQCGWFQFLA-UHFFFAOYNA-N
SMILES: O.O.O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)