missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sisomicin sulfate, 90%

CAS: 53179-09-2 | C19H39N5O11S | 545.605 g/mol

Supplier:  Thermo Scientific Chemicals J66063ME

 View more versions of this product

Catalog No. AAJ66063ME


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sisomicin sulfate
70°C to 73°C
>110°C (230°F)
C19H39N5O11S
500 mg
Soluble in water
CC1(COC(C(C1NC)O)OC2C(CC(C(C2O)OC3C(CC=C(O3)CN)N)N)N)O.OS(=O)(=O)O
545.605
692.72
Powder
53179-09-2
155°C
Plastic bottle
MFCD00058308
sisomicin sulfate
JVTNJDPXUPRGIE-DAVQCCERSA-N
(2S,3R,4S,5S)-2-[(2S,3S,4R,6S)-4,6-diamino-3-[[3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
123134107
90%

Chemical Identifiers

53179-09-2
545.605
JVTNJDPXUPRGIE-DAVQCCERSA-N
123134107
CC1(COC(C(C1NC)O)OC2C(CC(C(C2O)OC3C(CC=C(O3)CN)N)N)N)O.OS(=O)(=O)O
C19H39N5O11S
MFCD00058308
sisomicin sulfate
(2S,3R,4S,5S)-2-[(2S,3S,4R,6S)-4,6-diamino-3-[[3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H360

EINECSNumber : 258-414-4

RTECSNumber : WK2288000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only