missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sisomicin sulfate, 90%

Catalog No. p-7076641
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66063ME 500 mg
AAJ6606303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66063ME Supplier Thermo Scientific Chemicals Supplier No. J66063ME
Only null left

CAS: 53179-09-2 | C19H39N5O11S | 545.605 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53179-09-2
Molecular Formula C19H39N5O11S
Molecular Weight (g/mol) 545.605
MDL Number MFCD00058308
InChI Key JVTNJDPXUPRGIE-DAVQCCERSA-N
Synonym sisomicin sulfate
PubChem CID 123134107
IUPAC Name (2S,3R,4S,5S)-2-[(2S,3S,4R,6S)-4,6-diamino-3-[[3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMILES CC1(COC(C(C1NC)O)OC2C(CC(C(C2O)OC3C(CC=C(O3)CN)N)N)N)O.OS(=O)(=O)O

Specifications

Melting Point 70°C to 73°C
Boiling Point 155°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 500 mg
Solubility Information Soluble in water
Formula Weight 692.72
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Sisomicin sulfate
Hazard Category H360
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 258-414-4
RTECSNumber WK2288000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only