missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sirtinol

Catalog No. AAJ64486MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64486MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64486MA Supplier Thermo Scientific Chemicals Supplier No. J64486MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 410536-97-9 | C26H22N2O2, C26H22N2O2 | 394.47 g/mol

Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 410536-97-9
Molecular Formula C26H22N2O2
Synonym (E)-2-((2-Hydroxynaphthalen-1-yl)methyleneamino)-N-(1-phenylethyl)benzamide

Specifications

Quantity 10 mg
Description Off-white to Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases
Formula Weight 394.46g/mol
Solubility Soluble in water, DMSO and methanol
Product Type Signal Transduction Reagent
Form Crystals

RUO – Research Use Only