CAS RN 410536-97-9
IUPAC Name:
2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
Synonyms:
2-({[(1Z)-2-oxonaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
Molecular Weight (g/mol):
394.47
Molecular Formula:
C26H22N2O2
InChi Key:
YUGODMKHHCZZOI-ZVTCDHROSA-N
SMILES:
C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Sirtinol
CAS: 410536-97-9 Molecular Formula: C26H22N2O2,C26H22N2O2 Molecular Weight (g/mol): 394.47 InChI Key: YUGODMKHHCZZOI-ZVTCDHROSA-N Synonym: (E)-2-((2-Hydroxynaphthalen-1-yl)methyleneamino)-N-(1-phenylethyl)benzamide IUPAC Name: 2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide SMILES: C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1