missing translation for 'onlineSavingsMsg'
Learn More

(-)-Sinigrin hydrate, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014131
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014131 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014131 Supplier Merck Emd Millipore Supplier No. 0029010MG
Featured Industries: Food and Beverages

Specifications

CAS 3952-98-5
Linear Formula C10H16 KNO9S2 · xH2 O
MDL Number MFCD00149447
Melting Point 128°C
Type Analytical Standard
Quantity 10 mg
Formula Weight 397.46 (anhydrous basis)
Percent Purity ≥99% (TLC)
Grade Analytical Standard