missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs

Catalog No. 50400731
Change view
Click to view available options
Capacity (Metric):
86 mL
90 mL
92 mL
Sterility:
Non-sterile
Sterile
Channels:
1
384
96
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
50400731 86 mL Non-sterile 96 25/Cs.
14222419 92 mL Non-sterile 384 25/Cs.
14222426 86 mL Sterile 96 Individually Wrapped; 25/Cs.
50400722 90 mL Non-sterile 1 25/Cs.
14222420 92 mL Sterile 384 Individually Wrapped; 25/Cs.
14222414 90 mL Sterile 1 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50400731 Supplier Axygen™ Supplier No. RESSW96LP

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 86 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
Packaging 25/Cs.
Channels 96
Disposable Single-use