Learn More
Axygen™ Single Well Low Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Supplier:  Axygen™ RESSW1LP
 View more versions of this product
Catalog No. 50-400-722
Only null left
Explore available promotions
Description
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
Specifications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic
90 mL
Non-sterile
25/Cs.
25/Cs.
Single-use
Clear
Polypropylene
Reagent Reservoir
25/Pk.
1
Product Suggestions
Product Suggestions