Learn More
Axygen™ Single Well High Profile Reagent Reservoirs
Catalog No. 50400726
Change view
Click to view available options
Capacity (Metric):
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
12
384
8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels
50400726 290 mL Non-sterile 8
14222418 282 mL Sterile 384
14222422 290 mL Sterile 8
14222412 290 mL Sterile 12
50400720 290 mL Non-sterile 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50400726 Supplier Axygen™ Supplier No. RESSW8HP
Manufactured with virgin polypropylene for excellent chemical resistance
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
Specifications
Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 290 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Channels 8
Disposable Single-use
Show More Show Less