missing translation for 'onlineSavingsMsg'
Learn More

Sinapic acid Matrix Substance for MALDI-MS (T), ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111014480
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014480 5 g
111014479 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014480 Supplier Merck Emd Millipore Supplier No. 854295G

Sinapic acid is a type of phenolic acid. Removal of amino group from phenylalanine, catalysing with enzyme phenylalanine ammonia-lyase forms cinnamic acid. The precursor of hydroxycinnamates is produced after hydroxylation of benzene ring. One of the precursors produced is sinapic acid.

Specifications

CAS 530-59-6
Linear Formula C11H12O5
MDL Number MFCD00004401
Melting Point 202°C
Synonym 3,5-Dimethoxy-4-hydroxycinnamic acid; 4-Hydroxy-3,5-dimethoxy-cinnamic acid; Sinapinic acid
Solubility Soluble in dioxane
Type Matrix Substance for MALDI-MS
Quantity 5 g
Formula Weight 224.21
Percent Purity ≥99.0% (T)
Grade Matrix Substance for MALDI-MS
Show More Show Less