missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015563
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015563 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015563 Supplier Merck Emd Millipore Supplier No. PHR14381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.Simvastatin is a hypolipidemic drug, which lowers plasma cholesterol by inhibiting 3-hydroxy-3-methyl-glutaryl-coenzyme A (HMG-CoA) reductase. It is widely used in the treatment of hypercholesterolemia.

Specifications

CAS 79902-63-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C25H38O5
MDL Number MFCD00072007
Melting Point 127°C to 132°C
Synonym Simvastatin
Type Certified Reference Material
Quantity 1 g
Formula Weight 418.57
Grade Certified Reference Material