missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin, MilliporeSigma™ Supelco™

Catalog No. 111001883
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001883 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001883 Supplier Merck Emd Millipore Supplier No. Y0002069

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 79902-63-9
Linear Formula C25H38O5
MDL Number MFCD00072007
Melting Point 127°C to 132°C
Type Pharmacopeia and Metrological Institutes Standards
Quantity 5 mg
Formula Weight 418.57
Grade Pharmaceutical Primary Standard