missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific T100 BIOTUBE™ Tube Racks
T100 BIOTUBE™ Tube Racks are suitable for use in refrigerators, freezers or water baths. Ideal for freezing and storing samples.
$149.95 - $211.68
Specifications
| Holds | Microtubes (1.2 mL) |
|---|---|
| Material | Polypropylene |
| Positions | 96 |
| Quantity | 10/Cs. |
| Product Type | T100 BIOTUBE Tube Rack |
| Catalog Number | Mfr. No. | Color | Sterility | Includes | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Sterility | Includes | Price | Quantity & Availability | |||
|
05500
|
Simport™ Scientific
T1001B |
Blue | Non-sterile | 96 Plain Individual Tubes |
Case of 10 Each for $172.82
|
|
|||
|
055001
|
Simport™ Scientific
T1001G |
Green | Non-sterile | 96 Plain Individual Tubes |
Case of 10 Each for $172.82
|
|
|||
|
0550010
|
Simport™ Scientific
T100 3R |
Red | Non-sterile | 12 Strips of 8 Tubes |
Case of 10 Each for $152.66
|
|
|||
|
0550012
|
Simport™ Scientific
T1004B |
Blue | Sterile | 12 Strips of 8 Tubes |
Case of 10 Each for $161.50
|
|
|||
|
0550013
|
Simport™ Scientific
T1004G |
Green | Sterile | 12 Strips of 8 Tubes |
Case of 960 Each for $149.95
|
|
|||
|
0550015
|
Simport™ Scientific
T1004Y |
Yellow | Sterile | 12 Strips of 8 Tubes |
Case of 10 Each for $154.92
|
|
|||
|
0550014
|
Simport™ Scientific
T1004R |
Red | Sterile | 12 Strips of 8 Tubes |
Case of 10 Each for $149.95
|
|
|||
|
055004
|
Simport™ Scientific
T1002B |
Blue | Sterile | 96 Plain Individual Tubes |
Case of 960 Each for $194.74
|
|
|||
|
055005
|
Simport™ Scientific
T1002G |
Green | Sterile | 96 Plain Individual Tubes |
Case of 10 Each for $211.68
|
|
|||
|
055007
|
Simport™ Scientific
T1002Y |
Yellow | Sterile | 96 Plain Individual Tubes |
Case of 10 Each for $194.74
|
|
|||
|
055009
|
Simport™ Scientific
T100 3G |
Green | Non-sterile | 12 Strips of 8 Tubes |
Case of 10 Each for $152.66
|
|
|||
Description
- Stands on four legs and can be removed from the base of the box and placed on a lab counter as a self-standing support
- Can be placed in a refrigerator or freezer shelf for improved air circulation around tubes, or in a water bath to allow controlled warming of the tubes and their contents
- Standard 96-well on center spacing of tubes
- Color coding system using colored interchangeable plastic grids
- Easy to read numbers and letters used on the box cover for sample identification
- Also ideal for storing, freezing and transporting reagents and specimens
- Availaible in red, blue, green and yellow
Specifications
| Microtubes (1.2 mL) | |
| 96 | |
| T100 BIOTUBE Tube Rack |
| Polypropylene | |
| 10/Cs. |