Learn More
Simport™ Scientific T100 BIOTUBE™ Tube Racks
T100 BIOTUBE™ Tube Racks are suitable for use in refrigerators, freezers or water baths. Ideal for freezing and storing samples.
Supplier:  Simport™ Scientific T100 3G
 View more versions of this product
Catalog No. 055009
Only null left
Description
Description
  • Stands on four legs and can be removed from the base of the box and placed on a lab counter as a self-standing support
  • Can be placed in a refrigerator or freezer shelf for improved air circulation around tubes, or in a water bath to allow controlled warming of the tubes and their contents
  • Standard 96-well on center spacing of tubes
  • Color coding system using colored interchangeable plastic grids
  • Easy to read numbers and letters used on the box cover for sample identification
  • Also ideal for storing, freezing and transporting reagents and specimens
  • Availaible in red, blue, green and yellow
Specifications
Chemical Identifiers
Microtubes (1.2 mL)
96
T100 BIOTUBE Tube Rack
Polypropylene
10/Cs.
Specifications
Green
Polypropylene
Non-sterile
12 Strips of 8 Tubes
Microtubes (1.2 mL)
96
10/Cs.
T100 BIOTUBE Tube Rack
Product Suggestions
Product Suggestions