Learn More
Simport™ Scientific MultiRack™ Tube Racks
Compact, lightweight and stackable. MultiRack™ Tube Racks are composed of a chemical-resistant acetal polymer that won't shatter or stain.
 View more versions of this product
Catalog No. 05407113
Only null left
Includes: Choose from three models that hold up to 84 × 13mm diameter tubes; 60 × 16mm including 15mL centrifuge tubes; or units accepting up to 18 × 50mL centrifuge tubes (25 to 30mm diameter).
Description
Description
  • Three-tiered racks are stable in water baths
  • Base tier has rounded wells with drain holes
  • Interlocking feet allow stacking
  • With handles for portability
  • Available in 5 colors
  • Choose from three models that hold up to 84 x 13 mm diameter tubes; 60 x 16 mm including 15 mL centrifuge tubes; or units accepting up to 18 x 50 mL centrifuge tubes (25 to 30 mm diameter)
Specifications
Chemical Identifiers
Acetal Polymer
11.5 in.
2.5 in.
6.5 cm
29.3 cm
4.5 in.
Tube Rack
11.5 cm
Specifications
Orange
Acetal Polymer
15mL Centrifuge tubes and urine collection tubes
11.5 in.
2.5 in.
Tube Rack
11.5 cm
Tubes (25 to 30 mm)
18
29.3 cm
4.5 in.
30 mm
6.5 cm
Product Suggestions
Product Suggestions