missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Capinsert™ Cap Inserts, Assorted

Catalog No. 056705
Change view
Click to view available options
Color:
Assorted (Blue, Lilac, Red, Yellow, White)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
056705 Assorted (Blue, Lilac, Red, Yellow, White)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 056705

Made of polypropylene.

The Capinsert™ is used to color code a multitude of products such as tubes and vials. It is inserted on top of the closure and has a write-on frosted area for sample identification.

  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings

Order Info

Available in 11 colors; assorted packages contain 100 of each color

Specifications

Pre-sterilized No
Autoclavable Autoclavable
Material Polypropylene
Color Assorted (Blue, Lilac, Red, Yellow, White)
For Use With (Application) Used to color code and write on frosted area for sample identification
Sterility Non-sterile
Tube Material Polypropylene
For Use With (Equipment) Micrewtubes Microcentrifuge Tubes and Other Products