missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific Nonsterile Screw Caps
Description
- Made of polypropylene
- Available with or without silicone washer
- Long skirt and super fast thread design for easy seal
- Autoclavable
Specifications
Specifications
| Includes | Silicone washer |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Natural |
| Sterility | Non-sterile |
| For Use With (Equipment) | Sample Tubes |