missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SimpliAmp™ Thermal Cycler + Plastics Package, EnduraPlates

Catalog No. A39294
Encompass
Change view
Click to view available options
96-well 3 zones VeriFlex, EnduraPlates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A39294 96-well 3 zones VeriFlex, EnduraPlates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39294 Supplier Applied Biosystems™ Supplier No. A39294

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This SimpliAmp Thermal Cycler package includes a thermal cycler plus MicroAmp EnduraPlates and MicroAmp Clear Adhesive Films.

SimpliAmp Thermal Cycler
The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow. Innovations like a responsive color touch-screen and VeriFlex temperature control technology enable simple, accurate optimization. And now you can access your SimpliAmp cycler with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.

For detailed instrument information and specifications, see the SimpliAmp Thermal Cycler.

MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.

Key features:

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
  • Available in multiple colors for easy organization of laboratory assays
  • Excellent mechanical stability and flatness for your complete workflow needs

MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.

Key features:

  • Adhesive provides tight seal over wells to prevent leaks and evaporation.
  • Easy to apply and remove
  • Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)

Specifications

Format 96-well 3 Zones VeriFlex
Thermal Accuracy ±0.25°C (35°C to 99.9°C)
Temperature Range (Metric) 0 to 100°C
Content And Storage SimpliAmp Thermal Cycler, 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total), 1 x MicroAmp Clear Adhesive Films (100 films)
Thermal Uniformity <0.5 °C (20 s after reaching 95°C)
Dimensions 46.0 x 24.0 x 21.0 cm
Capacity 96-well 3 Zones VeriFlex
Voltage 100 to 240 V
Electrical Requirements 100 to 240 V, 50 to 60 Hz, 600 VA (Max.)
Display Type 8 in. Color TFT LCD
Product Type PCR Instrument
Height (English) 8.3 in.
Depth (English) 18.1 in.
Height (Metric) 21.0 cm
Width (Metric) 24 cm
Width (English) 9.5 in.
Depth (Metric) 46.0 cm
Thermal Range 0°C to 100°C
For Use With (Application) PCR
Frequency 50 to 60 Hz
Length (English) 18.1 in.
Ramp Rate 3°C/s
Length (Metric) 46.0 cm
User Interface 8 in. Color TFT LCD
Range 0 to 100.0 °C
Wattage 600 W (Max.)
Max. Ramp Rate 4°C/s
No. of Programs >1000
Memory USB and 2000 MB Onboard Memory; Onboard Capacity >1000 Protocols
Volume (Metric) 96-well, 0.2 mL VeriFlex Block
Weight 8.3 kg (18.3 lb.)
Block Configurations 96-well 3 Zones VeriFlex
Show More Show Less