missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SimpliAmp™ Thermal Cycler + Extended Warranty + Plastics Packages

Catalog No. A36970
Encompass
Change view
Click to view available options
Warranty:
5 years of total service coverage
5 years of total service coverage with instrument rapid exchange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Warranty
A36970 5 years of total service coverage
A36969 5 years of total service coverage with instrument rapid exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A36970 Supplier Applied Biosystems™ Supplier No. A36970
  • ONLINE EXCLUSIVE PRICE
    $10,325.23
    $6,711.40 / Each

    Save $3,613.83

    35.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.

The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow. Innovations like a responsive color touch-screen and Applied Biosystems VeriFlex temperature control technology enable simple, accurate optimization.

This package consists of a SimpliAmp Thermal Cycler, plastics consumables, and an extended warranty. You can access your SimpliAmp cycler with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.

Rapid Exchange (REX) extended warranty

With the REX extended warranty, if your instrument needs repair, just call our Support Center and we will exchange it for a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.

The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.

AB Repair Center (ABRC) extended warranty

With ABRC support, factory-trained engineers will perform maintenance or repair your instrument at our remote service center. Just send us your instrument and we will maintain or repair it and ship it back to you typically within three weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments that must maintain asset tag consistency to comply with regulated protocols.

Contact your Service and Support Representative for details at service.sales@thermofisher.com.

MicroAmp plastics

This package contains several MicroAmp plastic consumables to get you started. All of our plastic consumables are validated with Applied Biosystems instruments for optimal fit and performance. The following plastic consumables will be delivered along with your instrument when you order this package:

  • 1 x MicroAmp 8-Tube Strip with attached domed caps, 0.2 mL (Cat. No. A30589)
  • 1 x MicroAmp Clear Adhesive Film (Cat. No. 4306311)
  • 5 x MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates (Cat. No. A32811)
  • 1 x MicroAmp 32-Well Clear Adhesive Film (Cat. No. A32812)

Specifications

Content And Storage • SimpliAmp Thermal Cycler
• 3-year AB Repair Center (ABRC) extended warranty
• 1 x MicroAmp 8-Tube Strip with Attached Domed Caps, 0.2 mL (125 strips)
• 1 x MicroAmp Clear Adhesive Film (100 films)
• 5 x MicroAmp TriFlex 3 x 32-well PCR Reaction Plates (20 plates/pack, 100 plates total)
• 1 x MicroAmp 32-well Clear Adhesive Film (100 films)

High-throughput Compatibility Manual Protocols
Reaction Speed Fast, Standard
Dimensions 18.1 x 9.5 x 8.3 in. (46 x 24 x 21 cm)
Capacity 96-well, 0.2 mL
Voltage 100/240 V
Display Type 8 in. Color TFT LCD Touchscreen
Product Type Thermal Cycler
Warranty 5 years of total service coverage
Thermal Range 0°C to 100°C
User Interface USB, Wi-Fi, Ethernet
Max. Ramp Rate 4°C/sec. (Block), 3°C/sec. (Sample)
Quantity 1 Package
Weight 18.3 lb. (8.5 kg)
Block Configurations Veriflex Block
Show More Show Less
What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf