missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Trifluoromethanesulfonate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 256.93 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Silver Trifluoromethanesulfonate |
Safety and Handling
| TSCA | Yes |