accessibility menu, dialog, popup

UserName

CAS RN 2923-28-6

IUPAC Name: silver(1+) trifluoromethanesulfonate
Synonyms: silver triflate
Molecular Weight (g/mol): 256.93
Molecular Formula: CAgF3O3S
InChi Key: QRUBYZBWAOOHSV-UHFFFAOYSA-M
SMILES: [Ag+].[O-]S(=O)(=O)C(F)(F)F
Physical Form 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)