missing translation for 'onlineSavingsMsg'
Learn More

Silver Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T13311G
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13311G 1 g
T133110G 10 g
T133125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T13311G Supplier TCI America Supplier No. T13311G

Chemical Identifiers

CAS 2923-28-6
Molecular Formula CAgF3O3S
Molecular Weight (g/mol) 256.93
MDL Number MFCD00013226
InChI Key QRUBYZBWAOOHSV-UHFFFAOYSA-M
Synonym silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
PubChem CID 76223
IUPAC Name silver(1+) trifluoromethanesulfonate
SMILES [Ag+].[O-]S(=O)(=O)C(F)(F)F

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 256.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Silver Trifluoromethanesulfonate