missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfide, Premion™, 99.995% (metals basis)

CAS: 21548-73-2 | Ag2S | 247.80 g/mol

$308.93 - $1221.59

Chemical Identifiers

CAS 21548-73-2
Molecular Formula Ag2S
Molecular Weight (g/mol) 247.80
MDL Number MFCD00003406
InChI Key XUARKZBEFFVFRG-UHFFFAOYSA-N
IUPAC Name disilver(1+) sulfanediide
SMILES [S--].[Ag+].[Ag+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1211306
View Documents
Thermo Scientific Chemicals
01211306
5 g
Each for $308.93
Only null left
 
AA1211314
View Documents
Thermo Scientific Chemicals
01211314
25 g
Each for $1,221.59
Only null left
 
Description

Description

It finds use as a photosensitizer in photography. Blackening the silver for jewelleries and disinfection/sterilization of reservoirs. Anti-bacterial and anti-microbial agents. Laboratory reagents. A mixed ionic-electronic conductor. In IR detectors, photoconductors, photovoltaic cells, electrochemical storage cells, etc. Black colorant in ceramic glazes and for niello metal work. Autometallography in the detection of catalytic tissue metals and colloidal gold particles bound to macromolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21548-73-2
247.80
XUARKZBEFFVFRG-UHFFFAOYSA-N
[S--].[Ag+].[Ag+]
Ag2S
MFCD00003406
disilver(1+) sulfanediide

Specifications

825°C
21548-73-2
5 g
Ag2S
Insoluble in most of the solvents. Insoluble in acids,alkalies and aqueous ammoniums.
[S--].[Ag+].[Ag+]
247.80
Odorless
Dark grey to black powder/flake/crystalline/beads
Silver sulfide
Gray to Black
Powder
99.998% (metals basis)
MFCD00003406
XUARKZBEFFVFRG-UHFFFAOYSA-N
disilver(1+) sulfanediide
247.8
Light sensitive
7.317
Videos
Safety and Handling

Safety and Handling

H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 244-438-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only