accessibility menu, dialog, popup

UserName

AgAgS

CAS RN 21548-73-2

IUPAC Name: disilver(1+) sulfanediide
Synonyms: silver sulfide
Molecular Weight (g/mol): 247.8
Molecular Formula: Ag2S
InChi Key: XUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES: [S--].[Ag+].[Ag+]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)