missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, pure, 10 g/l solution in sulfuric acid

Catalog No. AC279480010
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279480010 1 L
AC279480025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279480010 Supplier Thermo Scientific Chemicals Supplier No. 279480010

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Silver sulfate
Color Colorless
Name Note 10g/L Solution in Sulfuric Acid
CAS 10294-26-5, 7664-93-9
Molecular Formula Ag2H2O4S
Linear Formula Ag2SO4
MDL Number MFCD00003407
Fieser 01,1015; 03,254; 04,435
Density 1.8400g/mL
Merck Index 15, 8668
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES [Ag+].[Ag+].OS(O)(=O)=O
IUPAC Name disilver(1+) sulfuric acid
Molecular Weight (g/mol) 313.81
PubChem CID 159865
Formula Weight 311.79
Grade Pure
Packaging Glass bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Show More Show Less

RUO – Research Use Only