missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfadiazine 98.0+%, TCI America™
$108.11 - $348.57
Chemical Identifiers
| CAS | 22199-08-2 |
|---|---|
| Molecular Formula | C10H9AgN4O2S |
| Molecular Weight (g/mol) | 357.136 |
| MDL Number | MFCD00072101 |
| InChI Key | UEJSSZHHYBHCEL-UHFFFAOYSA-N |
| Synonym | silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver |
| PubChem CID | 441244 |
| ChEBI | CHEBI:9142 |
| IUPAC Name | silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide |
| SMILES | C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] |
Chemical Identifiers
| 22199-08-2 | |
| 357.136 | |
| UEJSSZHHYBHCEL-UHFFFAOYSA-N | |
| 441244 | |
| silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide |
| C10H9AgN4O2S | |
| MFCD00072101 | |
| silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver | |
| CHEBI:9142 | |
| C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] |
Specifications
| 285°C | |
| 22199-08-2 | |
| 5 g | |
| MFCD00072101 | |
| UEJSSZHHYBHCEL-UHFFFAOYSA-N | |
| silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide | |
| 441244 | |
| 357.14 | |
| Silver Sulfadiazine |
| White-Yellow | |
| Crystalline Powder | |
| C10H9AgN4O2S | |
| silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver | |
| C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] | |
| 357.136 | |
| CHEBI:9142 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : WP1950000
TSCA : No