accessibility menu, dialog, popup

UserName

AgNH2SOONNN

CAS RN 22199-08-2

IUPAC Name: silver(1+) 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
Synonyms: silver sulfadiazine silver(1+) sulfadiazinate silvadene silver sulphadiazine SSD dermazin thermazene 磺胺嘧啶银 sulfadiazine silver salt ssd (1% silver sulfadiazine cream usp)
Molecular Weight (g/mol): 357.14
Molecular Formula: C10H9AgN4O2S
InChi Key: UEJSSZHHYBHCEL-UHFFFAOYSA-N
SMILES: [Ag+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)