Learn More
Silver Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$267.85 - $454.74
Chemical Identifiers
| CAS | 7440-22-4 |
|---|---|
| Molecular Formula | Ag |
| Molecular Weight (g/mol) | 107.87 |
| MDL Number | MFCD00003397 |
| InChI Key | BQCADISMDOOEFD-UHFFFAOYSA-N |
| PubChem CID | 23954 |
| ChEBI | CHEBI:9141 |
| IUPAC Name | silver |
| SMILES | [Ag] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLAG22Y
|
SPEX CertiPrep Group LLC
PLAG22Y |
125 mL | Triple-leached LDPE bottle |
Each for $267.85
|
|
||||
|
PLAG2-2X
|
SPEX CertiPrep Group LLC
PLAG22X |
500 mL | Triple-leached LDPE bottle |
Each for $454.74
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-22-4 | |
| 107.87 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| CHEBI:9141 | |
| [Ag] |
| Ag | |
| MFCD00003397 | |
| 23954 | |
| silver |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Ag | |
| AA, ICP | |
| Certified Reference Material | |
| [Ag] | |
| silver | |
| 125 mL | |
| CHEBI:9141 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLAG2-2Y, -2T, -2X 10,000 PLAG2-3Y, -3X |
| 7440-22-4,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00003397 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 107.87 | |
| 23954 | |
| Assurance | |
| Silver Standard |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only