missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, APS 1.3-3.2 micron, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol

Supplier:  Thermo Scientific Chemicals 04159918

 View more versions of this product

Catalog No. AA4159918


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver powder
50 g
APS 0.8 to 1.5 Micron
99.9%, metal basis
Ag
UN3077
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.9%
7440-22-4
Characteristic
Spherical Powder
(metals basis)
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.

Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only