missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, APS 1.3-3.2 micron, 99.9% (metals basis)

Catalog No. p-7051585
Change view
Click to view available options
Quantity:
10 g
250 g
50 g
Catalog No. Quantity
AA4159909 10 g
AA4159918 50 g
AA4159930 250 g
3 options

Catalog No. AA4159909

Supplier: Thermo Scientific Chemicals 04159909

Only null left

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 10 g
Form Spherical Powder
Assay Percent Range 99.9%, metal basis
UN Number UN3077
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver
Hazard Category Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
Hazard Statement Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only