missing translation for 'onlineSavingsMsg'
Learn More

Silver, plasma standard solution, Specpure™ Ag 1000μg/mL

CAS: 7440-22-4 | Ag | 107.87 g/mol

$139.29 - $139.29

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13849AE
View Documents
Thermo Scientific Chemicals
013849AE
100 mL
Each for $139.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
CHEBI:9141
[Ag]
Ag
MFCD00003397
23954
silver

Specifications

7440-22-4
Liquid
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
100 mL
CHEBI:9141
Silver, plasma standard solution
23 hPa (17mm Hg) at 20°C
Ag
Colorless
[Ag]
107.87
23954
Nitric acid: 5%; Silver: 0.1%; Water: 94.9%
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only