missing translation for 'onlineSavingsMsg'
Learn More

Silver Oxide, Powder, 99.9%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1090125GM

 View more versions of this product

Catalog No. 18613211


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

20667-12-3
125 g
Reagent
Amber Glass Bottle
1
99.9%
0.01

Chemical Identifiers

20667-12-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.