missing translation for 'onlineSavingsMsg'
Learn More

Silver Oxide, Powder, 99.9%, Spectrum™ Chemical

Catalog No. p-7166275
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18613209 25 g Amber Glass Bottle
18613210 100 g Amber Glass Bottle
18613211 125 g Amber Glass Bottle
18613212 500 g Amber Glass Bottle
4 options

Catalog No. 18613209

Supplier: Spectrum Chemical Mfg Cor S109025GM

Only null left

Chemical Identifiers

CAS 20667-12-3

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99.9%
Grade Reagent
Loss on Drying 0.01
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.