missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrate solution, Volumetric, Reag. Ph. Eur., 0.1Â M AgNO3 (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1Â M AgNO3 (0.1N)
$250.68 - $2337.50
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6001333
|
Honeywell Chemicals
35375500ML |
500 mL |
Each for $250.68
|
|
|||||
|
6001331
|
Honeywell Chemicals
353751L |
1 L |
Each for $396.33
|
|
|||||
|
6001332
|
Honeywell Chemicals
3537525L |
2.5 L |
Each for $762.30
|
|
|||||
|
6001334
|
Honeywell Chemicals
353755LVP |
5 L |
Each for $1,673.60
|
|
|||||
|
60046579
|
Honeywell Chemicals
353756X1L |
6 x 1 L |
Case of 6 Each for $1,988.63
|
|
|||||
|
6001330
|
Honeywell Chemicals
3537510LVP |
10 L |
Each for $2,337.50
|
|
|||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| [Ag+].[O-][N+]([O-])=O |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 |
Specifications
| 7761-88-8 | |
| AgNO3 | |
| UN3082 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87g/mol | |
| Silver nitrate solution |
| 500 mL | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| 0.1M |
Safety and Handling
P273-P280-P305 + P351 + P338-P337 + P313-P391-P501