missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate solution, 0.1N, TitriPUR™, MilliporeSigma™

Catalog No. M1090811000
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1090811000 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1090811000 Supplier Technidata Supplier No. 1.09081.1000
Only null left

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Solubility Information Soluble (20°C)
Packaging Plastic Bottle
Density 1.01g/cm3 (20°C)
Chemical Name or Material Titripur™
Hazard Category SignalWord: Warning
Recommended Storage Store at +15°C to +25°C