missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, puriss. p.a., Honeywell Fluka™

Catalog No. 6001325
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001325 50 g
6001323 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001325 Supplier Honeywell Chemicals Supplier No. 8522850G
Only null left

Puriss. p.a.,≥99.5% (AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 50 g
Linear Formula AgNO3
UN Number UN1493
Formula Weight 169.87g/mol
Percent Purity ≥99.5% (AT)
Grade puriss. p.a.
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
RTECSNumber VW4725000