missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7072040
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001322 25 g
6001321 250 g
6001320 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001322 Supplier Honeywell Chemicals Supplier No. 3163025G
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 25 g
Linear Formula AgNO3
UN Number 1493
Formula Weight 169.87g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314-H410
EINECSNumber 231-853-9
RTECSNumber VW4725000