missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, Premion™, 99.9995% (metals basis)

Catalog No. AA1085822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1085822 100 g
AA1085806 5 g
AA1085814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1085822 Supplier Thermo Scientific Chemicals Supplier No. 01085822

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate finds use in silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain and staining in histology. It is also used as a stain in scanning electron microscopy and a key ingredient in several silver-based pharmaceutical compositions through appropriate manufacturing processes. Silver nitrate can be used to separate mixtures of alkenes by selective absorption. It is also used for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9995% (metals basis)
Solubility Information Freely soluble in water,alcohol,and aqueous ammonia. Slightly soluble in ether.
Formula Weight 169.87
Odor Odorless
Sensitivity Light sensitive
Appearance White crystals
Density 4.352
Chemical Name or Material Silver nitrate
Show More Show Less
Hazard Category H272-H290-H314-H335-H373
Precautionary Statement P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 231-853-9
RTECSNumber VW4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only