missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min

Catalog No. AA4308706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4308706 5 g
AA4308714 25 g
AA4308722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4308706 Supplier Thermo Scientific Chemicals Supplier No. 04308706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C
Color White
Physical Form Crystalline
Quantity 5 g
UN Number UN1479
Merck Index 14,8518
Solubility Information Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether.
Formula Weight 169.87
Percent Purity 99.995%
Grade Premion
Concentration or Composition (by Analyte or Components) Ag 63% min
Odor Odorless
Sensitivity Light sensitive
Density 4.352 g/mL
Assay (metals basis)
Chemical Name or Material Silver nitrate
Show More Show Less
Hazard Category H272-H290-H314-H335-H373
Hazard Statement GHS H Statement
H272-H314-H290
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Precautionary Statement P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 231-853-9
RTECSNumber VW4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only