missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, in Methanol, Certified, 0.0025N ±0.0001N (0.0025M), LabChem™

Catalog No. p-5388216
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC227422 1 L
LC227424 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC227422 Supplier LabChem Supplier No. LC227422
Only null left

Chemical Identifiers

CAS 7761-88-8 , 67-56-1
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 65°C
Melting Point -98°C
Color Colorless to Yellow
CAS Max % 99.96, 0.04
Physical Form Liquid
Quantity 1 L
Linear Formula AgNO3
UN Number UN1993
Flash Point 12°C
Solubility Information Soluble in water
Formula Weight 169.87
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0025N ±0.0001N (0.0025M)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 0.8g/cm3
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Density 0.8g/cm3
Chemical Name or Material Silver Nitrate, in Methanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up a cool, well ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature