missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, Honeywell Fluka™
Meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%
Supplier: Honeywell-Fluka 10220500G
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Silver Nitrate | |
| 7761-88-8 | |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87g/mol | |
| Vapor Density: 5.8 (vs air) |
| Meets analytical specification of Ph. Eur. BP, USP | |
| 500 g | |
| AgNO3 | |
| 1493 | |
| Soluble in ethanol, 20.8g/L, Soluble in benzene, 2.2g/L, Soluble in methanol, 35g/L | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| 99.8 to 100.5% |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H314-H410
EINECSNumber : 231-853-9
RTECSNumber : VW4725000