missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-33493
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S18125 25 g Glass Bottle
S181100 100 g Glass Bottle
S181500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S18125 Supplier Thermo Fisher Scientific Supplier No. FLS18125
Edge

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C
Melting Point 212°C
Color White
pH 5.4 to 6.4
Physical Form Solid
Quantity 25 g
Assay Percent Range Assay: ≥99.7%
Formula Weight 169.87
Percent Purity ≥99.7%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Silver Nitrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1: Oxidizer