missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Crystal, USP, 99.8 -100.5%, Spectrum™ Chemical

Catalog No. 18606237
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606237 125 g Amber Glass Bottle
18606239 250 g Amber Glass Bottle
18606238 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606237 Supplier Spectrum Chemical Mfg Cor Supplier No. SI115125GM
Only null left

SI115, 7761-88-8, AgNO3

Spectrum™ Chemical Silver Nitrate, Crystal, USP is used as an antibiotic for indolent wounds, corn and callus removal and as a Gonorrheal ophthalmia neonatorum prophylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99.8 to 100.5%
Grade USP
Packaging Amber Glass Bottle